Assay Details
Target Gene Details
Entrez Gene ID: | 6511 |
Gene Name: | solute carrier family 1 member 6 |
Gene Aliases: |
EAAT4 |
Location: |
Chr.19:14950033-15010643 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC1A6 | NM_005071.2 | 9 | 1568 | NP_005062.1 |
| NR_073589.1 | 9 | 1521 | ||
| XM_006722842.1 | 10 | 1751 | XP_006722905.1 | |
| XM_006722844.1 | 10 | 1644 | XP_006722907.1 | |
| XM_017027152.1 | 10 | 1839 | XP_016882641.1 | |
| AK302380.1 | 9 | 1498 | ||
| AK312996.1 | 9 | 1600 | ||
| HY352099.1 | 1 | 325 | ||
| U18244.1 | 9 | 1568 | AAA75314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063744 | Chr.19:14906349 - 14999775 on Build GRCh38 | Gain |
|
| nsv510756 | Chr.19:14926456 - 14952143 on Build GRCh38 | Deletion |
|
| nsv2424 | Chr.19:14934808 - 14978815 on Build GRCh38 | Deletion |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map