Assay Details
Target Gene Details
Entrez Gene ID: | 151742 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1L |
Gene Aliases: |
PP2C-epsilon, PP2CE, PPM1-LIKE |
Location: |
Chr.3:160755532-161078907 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1L | NM_139245.3 | 1 | 412 | NP_640338.2 |
| XM_011512440.2 | 1 | 1084 | XP_011510742.1 | |
| AY337264.1 | 1 | 408 | AAR00269.1 | |
| DB032505.1 | 1 | 412 |
More Information
Set Membership: |
|

Genomic Map