Assay Details
Target Gene Details
Entrez Gene ID: | 347148 |
Gene Name: | pyroglutamylated RFamide peptide |
Gene Aliases: |
26RFa, P518 |
Location: |
Chr.9:130893428-130896812 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRFP | NM_198180.2 | 3 | 861 | NP_937823.1 |
| AB109625.1 | 1 | 340 | BAC98934.1 | |
| BC101127.2 | 1 | 340 | AAI01128.1 | |
| BC101128.1 | 1 | 340 | AAI01129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621866 | Chr.9:130695968 - 131205700 on Build GRCh38 | Gain |
|
| nsv951789 | Chr.9:130880714 - 130963813 on Build GRCh38 | Deletion |
|
| nsv469903 | Chr.9:130890269 - 130908179 on Build GRCh38 | Loss |
|
| esv3621869 | Chr.9:130876741 - 130992658 on Build GRCh38 | Loss |
|
| esv2759717 | Chr.9:130862179 - 131153174 on Build GRCh38 | Loss |
|
| nsv6735 | Chr.9:130877388 - 130921852 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map