Assay Details
Target Gene Details
Entrez Gene ID: | 24141 |
Gene Name: | lysosomal associated membrane protein family member 5 |
Gene Aliases: |
BAD-LAMP, BADLAMP, C20orf103, LAMP-5, UNC-46 |
Location: |
Chr.20:9514358-9530524 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMP5 | NM_001199897.1 | 5 | 1643 | NP_001186826.1 |
| NM_012261.3 | 6 | 1775 | NP_036393.1 | |
| AL121740.1 | 6 | 1482 | CAB57330.1 | |
| BC050727.1 | 6 | 1478 | AAH50727.1 |
More Information
Set Membership: |
|

Genomic Map