Assay Details
Target Gene Details
Entrez Gene ID: | 5648 |
Gene Name: | mannan binding lectin serine peptidase 1 |
Gene Aliases: |
3MC1, CRARF, CRARF1, MAP1, MASP, MASP3, MAp44, PRSS5, RaRF |
Location: |
Chr.3:187216085-187292220 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MASP1 | NM_001879.5 | 16 | 4046 | NP_001870.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014596 | Chr.3:187106902 - 187410668 on Build GRCh38 | Gain |
|
| esv23875 | Chr.3:187188590 - 187250892 on Build GRCh38 | Gain |
|
| nsv508264 | Chr.3:187169641 - 187226709 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map