Assay Details
Target Gene Details
Entrez Gene ID: | 10767 |
Gene Name: | HBS1 like translational GTPase |
Gene Aliases: |
EF-1a, ERFS, HBS1, HSPC276, eRF3c |
Location: |
Chr.6:134960378-135054898 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HBS1L | NM_001145158.1 | 17 | 4876 | NP_001138630.1 |
| NM_006620.3 | 18 | 5002 | NP_006611.1 | |
| AB028961.1 | 13 | 4231 | BAA82990.1 | |
| AJ459826.1 | 18 | 4988 | CAD30873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604723 | Chr.6:134914396 - 135161428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map