Assay Details
Target Gene Details
Entrez Gene ID: | 84171 |
Gene Name: | lysyl oxidase like 4 |
Gene Aliases: |
LOXC |
Location: |
Chr.10:98247686-98268250 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL4 | NM_032211.6 | 4 | 733 | NP_115587.6 |
| XM_005270216.1 | 4 | 706 | XP_005270273.1 | |
| XM_006718020.1 | 5 | 896 | XP_006718083.1 | |
| AF338441.1 | 3 | 582 | AAK71934.1 | |
| AF395336.1 | 3 | 582 | AAL27543.1 | |
| AK172781.1 | 4 | 676 | BAD18762.1 | |
| AY036093.1 | 3 | 588 | AAK64186.1 | |
| BC013153.1 | 4 | 733 | AAH13153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831954 | Chr.10:98172815 - 98361277 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map