Assay Details
Target Gene Details
Entrez Gene ID: | 54456 |
Gene Name: | Mov10 RISC complex RNA helicase like 1 |
Gene Aliases: |
CHAMP, DJ402G11.8 |
Location: |
Chr.22:50090006-50161705 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOV10L1 | NM_001164104.1 | 2 | 228 | NP_001157576.1 |
| NM_001164105.1 | 2 | 305 | NP_001157577.1 | |
| NM_018995.2 | 2 | 228 | NP_061868.1 | |
| XM_005261923.3 | 2 | 506 | XP_005261980.1 | |
| XM_011530696.1 | 2 | 228 | XP_011528998.1 | |
| XM_011530697.1 | 2 | 506 | XP_011528999.1 | |
| XM_011530698.1 | 2 | 228 | XP_011529000.1 | |
| XM_011530699.1 | 2 | 190 | XP_011529001.1 | |
| XM_011530700.2 | 2 | 573 | XP_011529002.1 | |
| XM_011530701.1 | 2 | 228 | XP_011529003.1 | |
| XM_011530702.1 | 2 | 161 | XP_011529004.1 | |
| XM_011530704.1 | 2 | 228 | XP_011529006.1 | |
| XM_017028833.1 | 2 | 228 | XP_016884322.1 | |
| XM_017028834.1 | 2 | 228 | XP_016884323.1 | |
| XM_017028837.1 | 2 | 228 | XP_016884326.1 | |
| AF285604.1 | 2 | 172 | AAK31983.1 | |
| AK302401.1 | 2 | 305 | ||
| BC029552.1 | 2 | 508 | AAH29552.1 | |
| BC042828.1 | 2 | 508 | AAH42828.1 | |
| BC140944.1 | 3 | 229 | ||
| BC150137.1 | 2 | 170 | ||
| BC152539.1 | 2 | 170 | ||
| CR456466.1 | 2 | 151 | CAG30352.1 | |
| DB451595.2 | 2 | 228 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv582n27 | Chr.22:50024802 - 50157413 on Build GRCh38 | Gain |
|
| nsv1067062 | Chr.22:50089389 - 50130477 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map