Assay Details
Target Gene Details
Entrez Gene ID: | 51267 |
Gene Name: | C-type lectin domain family 1 member A |
Gene Aliases: |
CLEC-1, CLEC1 |
Location: |
Chr.12:10069504-10099065 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC1A | NM_001297748.1 | NP_001284677.1 | ||
| NM_001297749.1 | NP_001284678.1 | |||
| NM_001297750.1 | NP_001284679.1 | |||
| NM_001297751.1 | NP_001284680.1 | |||
| NM_016511.3 | NP_057595.2 | |||
| XM_011520687.2 | XP_011518989.1 | |||
| AF200949.1 | AAF36830.1 | |||
| AK075114.1 | BAC11410.1 | |||
| AK091989.1 | ||||
| AK291584.1 | ||||
| AK300979.1 | ||||
| AK313589.1 | ||||
| AY358587.1 | AAQ88950.1 | |||
| BC039072.1 | AAH39072.1 | |||
| BC067746.1 | AAH67746.1 | |||
| BQ722745.1 | ||||
| BX339075.2 | ||||
| BX380212.2 |
More Information
Set Membership: |
|

Genomic Map