Assay Details
Target Gene Details
Entrez Gene ID: | 7125 |
Gene Name: | troponin C2, fast skeletal type |
Gene Aliases: |
- |
Location: |
Chr.20:45823214-45833329 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNNC2 | NM_003279.2 | NP_003270.1 | ||
| XM_011529031.2 | XP_011527333.1 | |||
| AK291323.1 | ||||
| BC005323.1 | AAH05323.1 | |||
| BU854061.1 | ||||
| CR541864.1 | CAG46662.1 | |||
| CR541884.1 | CAG46682.1 | |||
| M22307.1 | AAA91854.1 | |||
| X07898.1 | CAA30737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
| nsv586132 | Chr.20:45785138 - 45842587 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs556074291] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map