Assay Details
Target Gene Details
Entrez Gene ID: | 1915 |
Gene Name: | eukaryotic translation elongation factor 1 alpha 1 |
Gene Aliases: |
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1 |
Location: |
Chr.6:73515750-73521032 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1A1 | NM_001402.5 | NP_001393.1 | ||
| XM_011535514.2 | XP_011533816.1 | |||
| AF174496.1 | AAF36537.1 | |||
| AF397403.1 | AAK93966.1 | |||
| AJ420488.1 | ||||
| AK055058.1 | ||||
| AK058051.1 | ||||
| AK058082.1 | ||||
| AK058163.1 | ||||
| AK098510.1 | ||||
| AK098518.1 | ||||
| AK098623.1 | ||||
| AK127384.1 | ||||
| AK289374.1 | ||||
| AK289442.1 | ||||
| AK290691.1 | ||||
| AK292639.1 | ||||
| AK297878.1 | ||||
| AK300922.1 | ||||
| AK304214.1 | ||||
| AK315827.1 | ||||
| AY043301.1 | AAK95378.1 | |||
| EF219140.1 | ||||
| EF362804.1 | ||||
| X03558.1 | CAA27245.1 | |||
| X16869.1 | CAA34756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763576 | Chr.6:73487915 - 73567526 on Build GRCh38 | Gain |
|
| esv2675873 | Chr.6:73518872 - 73519851 on Build GRCh38 | Deletion |
|
| nsv981157 | Chr.6:73517348 - 73519647 on Build GRCh38 | Duplication |
|
| nsv5350 | Chr.6:73512272 - 73557365 on Build GRCh38 | Deletion |
|
| esv3609512 | Chr.6:73518814 - 73519818 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map