Assay Details
Target Gene Details
Entrez Gene ID: | 10140 |
Gene Name: | transducer of ERBB2, 1 |
Gene Aliases: |
APRO5, APRO6, PIG49, TOB, TROB, TROB1 |
Location: |
Chr.17:50862223-50867978 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOB1 | NM_001243877.1 | 3 | 1693 | NP_001230806.1 |
| NM_001243885.1 | 2 | 1504 | NP_001230814.1 | |
| NM_005749.3 | 2 | 1707 | NP_005740.1 | |
| BC031406.1 | 1 | 1271 | AAH31406.1 |
More Information
Set Membership: |
|

Genomic Map