Assay Details
Target Gene Details
Entrez Gene ID: | 26207 |
Gene Name: | phosphatidylinositol transfer protein, cytoplasmic 1 |
Gene Aliases: |
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1 |
Location: |
Chr.17:67377281-67697263 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITPNC1 | NM_012417.3 | 1 | 612 | NP_036549.2 |
| NM_181671.2 | 1 | 612 | NP_858057.1 | |
| AK290838.1 | 1 | 434 | ||
| BC007905.2 | 1 | 85 | AAH07905.1 | |
| BC067095.1 | 1 | 333 | AAH67095.1 | |
| BG504645.1 | 1 | 613 |
More Information
Set Membership: |
|

Genomic Map