Assay Details
Target Gene Details
Entrez Gene ID: | 25862 |
Gene Name: | ubiquitin specific peptidase 49 |
Gene Aliases: |
- |
Location: |
Chr.6:41789896-41895361 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP49 | NM_001286554.1 | NP_001273483.1 | ||
| NM_018561.4 | 7 | 2200 | NP_061031.2 | |
| AJ586139.1 | CAE51939.1 | |||
| BC014176.2 | 7 | 2200 | AAH14176.1 | |
| DN992189.1 |
More Information
Set Membership: |
|

Genomic Map