Assay Details
Target Gene Details
Entrez Gene ID: | 3816 |
Gene Name: | kallikrein 1 |
Gene Aliases: |
KLKR, Klk6, hK1 |
Location: |
Chr.19:50819146-50823787 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK1 | NM_002257.3 | NP_002248.1 | ||
| XM_011526942.1 | XP_011525244.1 | |||
| AK293363.1 | ||||
| AY429508.1 | AAR10467.1 | |||
| AY703451.1 | AAU12569.1 | |||
| BC005313.1 | AAH05313.1 | |||
| BP323011.1 | ||||
| BT007253.1 | AAP35917.1 | |||
| M12706.1 | AAA59201.1 | |||
| M25629.1 | AAA36136.1 | |||
| X13561.1 | CAA31912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv204n21 | Chr.19:50817827 - 50832941 on Build GRCh38 | Loss |
|
| esv3583464 | Chr.19:50818135 - 50842312 on Build GRCh38 | Loss |
|
| dgv203n21 | Chr.19:50797973 - 50832687 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map