Assay Details
Target Gene Details
Entrez Gene ID: | 11222 |
Gene Name: | mitochondrial ribosomal protein L3 |
Gene Aliases: |
COXPD9, MRL3, RPML3 |
Location: |
Chr.3:131462201-131503016 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL3 | NM_007208.3 | NP_009139.1 | ||
| AK296626.1 | ||||
| AK301377.1 | ||||
| BC003375.2 | AAH03375.1 | |||
| CR456720.1 | CAG33001.1 | |||
| X06323.1 | CAA29639.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597781 | Chr.3:131272356 - 131472358 on Build GRCh38 | Gain |
|
| nsv829724 | Chr.3:131366925 - 131557541 on Build GRCh38 | Gain |
|
| nsv1006656 | Chr.3:131383932 - 132145572 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map