Assay Details
Target Gene Details
Entrez Gene ID: | 153222 |
Gene Name: | CREB3 regulatory factor |
Gene Aliases: |
C5orf41, LRF |
Location: |
Chr.5:173056349-173139288 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREBRF | NM_001168393.1 | NP_001161865.1 | ||
| NM_001168394.1 | NP_001161866.1 | |||
| NM_153607.2 | NP_705835.2 | |||
| XM_005265821.3 | XP_005265878.1 | |||
| XM_011534442.1 | XP_011532744.1 | |||
| AK124117.1 | ||||
| AK291677.1 | ||||
| AK300043.1 | ||||
| AY139008.1 | AAN28956.1 | |||
| AY174896.1 | AAO18732.1 | |||
| BC041709.1 | AAH41709.1 | |||
| BX647573.1 | CAI46104.1 | |||
| BX647768.1 | CAI46039.1 |
More Information
Set Membership: |
|

Genomic Map