Assay Details
Target Gene Details
Entrez Gene ID: | 145567 |
Gene Name: | tetratricopeptide repeat domain 7B |
Gene Aliases: |
TTC7L1, c14_5685 |
Location: |
Chr.14:90540588-90816479 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC7B | NM_001010854.1 | 20 | 2810 | NP_001010854.1 |
| NM_001320421.1 | 21 | 2857 | NP_001307350.1 | |
| XM_005267367.1 | 18 | 2334 | XP_005267424.1 | |
| XM_011536497.1 | 22 | 3081 | XP_011534799.1 | |
| XM_017021044.1 | 22 | 2987 | XP_016876533.1 | |
| XM_017021045.1 | 21 | 2825 | XP_016876534.1 | |
| XM_017021046.1 | 20 | 2868 | XP_016876535.1 | |
| XM_017021047.1 | 19 | 2485 | XP_016876536.1 | |
| AK126571.1 | 11 | 1543 | ||
| AL832848.1 | 19 | 2487 | CAI46128.1 | |
| BC035865.2 | 15 | 2106 | AAH35865.1 | |
| BC048270.2 | 14 | 1911 | AAH48270.2 | |
| BX247966.1 | 7 | 1149 | CAD62305.1 | |
| BX248275.1 | 16 | 2102 | CAD62603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520068 | Chr.14:90502030 - 90560888 on Build GRCh38 | Loss |
|
| nsv470654 | Chr.14:90489716 - 90561263 on Build GRCh38 | Loss |
|
| nsv456392 | Chr.14:90508674 - 90561264 on Build GRCh38 | Loss |
|
| esv3635309 | Chr.14:90382200 - 90607667 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs199994385] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map