Assay Details
Target Gene Details
Entrez Gene ID: | 84549 |
Gene Name: | MAK16 homolog |
Gene Aliases: |
MAK16L, RBM13 |
Location: |
Chr.8:33485167-33501260 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAK16 | NM_032509.3 | 10 | 3219 | NP_115898.2 |
| AF251062.1 | 10 | 3194 | AAK34952.1 | |
| AK225476.1 | 10 | 1189 | ||
| AL049422.1 | 1 | 2052 | ||
| BC028230.1 | 10 | 3187 | AAH28230.1 | |
| BC039740.1 | 10 | 3200 | AAH39740.1 | |
| BC050528.1 | 10 | 3187 | AAH50528.1 |
Target Gene Details
Entrez Gene ID: | 80185 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
C8orf41, MRT39 |
Location: |
Chr.8:33498509-33513601 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTI2 | NM_001102401.2 | NP_001095871.1 | ||
| NM_001265581.1 | NP_001252510.1 | |||
| NM_025115.3 | NP_079391.2 | |||
| XM_011544665.2 | XP_011542967.1 | |||
| AK026916.1 | BAB15590.1 | |||
| BC007387.2 | AAH07387.1 | |||
| BC066935.1 | AAH66935.1 | |||
| BX648410.1 |
More Information
Set Membership: |
|

Genomic Map