Assay Details
Target Gene Details
Entrez Gene ID: | 1521 |
Gene Name: | cathepsin W |
Gene Aliases: |
LYPN |
Location: |
Chr.11:65879813-65883741 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSW | NM_001335.3 | NP_001326.2 | ||
| AF013611.1 | AAB82449.1 | |||
| BC048255.1 | AAH48255.1 | |||
| BI833187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| nsv951019 | Chr.11:65874630 - 65899129 on Build GRCh38 | Deletion |
|
| nsv555215 | Chr.11:65814108 - 65909045 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map