Assay Details
Target Gene Details
Entrez Gene ID: | 2982 |
Gene Name: | guanylate cyclase 1 soluble subunit alpha |
Gene Aliases: |
GC-SA3, GUC1A3, GUCA3, GUCSA3, GUCY1A1, MYMY6 |
Location: |
Chr.4:155666710-155737062 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY1A3 | NM_000856.5 | NP_000847.2 | ||
| NM_001130682.2 | NP_001124154.1 | |||
| NM_001130683.3 | NP_001124155.1 | |||
| NM_001130685.2 | NP_001124157.1 | |||
| NM_001130687.2 | NP_001124159.1 | |||
| NM_001256449.1 | NP_001243378.1 | |||
| XM_005262955.2 | XP_005263012.1 | |||
| XM_005262956.2 | XP_005263013.1 | |||
| XM_005262957.2 | XP_005263014.1 | |||
| XM_006714196.2 | XP_006714259.1 | |||
| AK096588.1 | ||||
| AK309950.1 | ||||
| AK312952.1 | ||||
| DA518091.1 | ||||
| DA793192.1 | ||||
| DA908917.1 | ||||
| X66534.1 | CAA47145.1 | |||
| Y15723.1 | CAA75738.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437445 | Chr.4:155663245 - 155693802 on Build GRCh38 | Loss |
|
| esv2760878 | Chr.4:155640759 - 155680884 on Build GRCh38 | Loss |
|
| nsv1022201 | Chr.4:155657250 - 155676831 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map