Assay Details
Target Gene Details
Entrez Gene ID: | 4207 |
Gene Name: | BORCS8-MEF2B readthrough |
Gene Aliases: |
LOC729991-MEF2B, MEF2B, MEF2BNB-MEF2B |
Location: |
Chr.19:19145567-19192591 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORCS8-MEF2B | NM_005919.3 | NP_005910.1 | ||
| NR_027307.1 | ||||
| NR_027308.1 | ||||
| AK301086.1 | ||||
| AK316328.1 | ||||
| BC126245.1 | ||||
| BC136457.1 | ||||
| BC171767.1 | ||||
| X63380.1 | CAA44978.1 | |||
| X68502.1 | CAA48515.1 |
Target Gene Details
Entrez Gene ID: | 100271849 |
Gene Name: | myocyte enhancer factor 2B |
Gene Aliases: |
RSRFR2 |
Location: |
Chr.19:19145567-19170289 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEF2B | NM_001145785.1 | NP_001139257.1 | ||
| AK057210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3556145 | Chr.19:19147168 - 19147525 on Build GRCh38 | Deletion |
|
| nsv819543 | Chr.19:19146335 - 19147548 on Build GRCh38 | Gain |
|
| nsv833779 | Chr.19:19052683 - 19202282 on Build GRCh38 | Loss |
|
| esv5981 | Chr.19:19147087 - 19147607 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| esv3093790 | Chr.19:19147213 - 19147487 on Build GRCh38 | Deletion |
|
| esv3643846 | Chr.19:19147203 - 19147482 on Build GRCh38 | Loss |
|
| esv2677504 | Chr.19:19147209 - 19147483 on Build GRCh38 | Deletion |
|
| esv2718291 | Chr.19:19147206 - 19147475 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map