Assay Details
Target Gene Details
Entrez Gene ID: | 64794 |
Gene Name: | DEAD-box helicase 31 |
Gene Aliases: |
PPP1R25 |
Location: |
Chr.9:132592997-132670403 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX31 | NM_001322340.1 | 18 | 2414 | NP_001309269.1 |
| NM_001322341.1 | 21 | 2737 | NP_001309270.1 | |
| NM_001322342.1 | 20 | 2137 | NP_001309271.1 | |
| NM_001322343.1 | 20 | 2196 | NP_001309272.1 | |
| NM_022779.8 | 20 | 2633 | NP_073616.6 | |
| NR_136309.1 | 19 | 2496 | ||
| XM_005272207.4 | 20 | 2320 | XP_005272264.1 | |
| XM_006717236.3 | 18 | 2441 | XP_006717299.1 | |
| AF335568.1 | 20 | 2634 | AAQ14889.1 | |
| AF335569.1 | 18 | 2415 | AAQ14890.1 | |
| AF427339.1 | 20 | 2634 | AAL26549.1 | |
| AK023695.1 | 16 | 1706 | BAB14644.1 | |
| AK027002.1 | 20 | 2217 | BAB15620.1 | |
| AK225121.1 | 20 | 2188 | ||
| AK225350.1 | 20 | 2218 | ||
| AK302670.1 | 20 | 2370 | ||
| BC012726.2 | 20 | 2259 | AAH12726.2 |
More Information
Set Membership: |
|

Genomic Map