Assay Details
Target Gene Details
Entrez Gene ID: | 101928847 |
Gene Name: | uncharacterized LOC101928847 |
Gene Aliases: |
- |
Location: |
Chr.11:112959279-112963460 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928847 | NR_120563.1 | |||
| AL833634.1 |
Target Gene Details
Entrez Gene ID: | 4684 |
Gene Name: | neural cell adhesion molecule 1 |
Gene Aliases: |
CD56, MSK39, NCAM |
Location: |
Chr.11:112961247-113278436 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAM1 | NM_000615.6 | 1 | 227 | NP_000606.3 |
| NM_001076682.3 | 1 | 227 | NP_001070150.1 | |
| NM_001242607.1 | 1 | 227 | NP_001229536.1 | |
| NM_001242608.1 | 1 | 227 | NP_001229537.1 | |
| NM_181351.4 | 1 | 227 | NP_851996.2 | |
| AB209443.1 | 1 | 106 | BAD92680.1 | |
| AF005070.1 | 1 | 72 | AAB61615.1 | |
| AK314589.1 | 1 | 199 | ||
| BC029119.1 | ||||
| BC047244.1 | AAH47244.1 | |||
| DA231540.1 | 1 | 198 | ||
| DA337649.1 | 1 | 201 | ||
| S71824.1 | AAB31836.1 | |||
| X16841.1 | CAA34739.1 |
More Information
Set Membership: |
|

Genomic Map