Assay Details
Target Gene Details
Entrez Gene ID: | 10930 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 2 |
Gene Aliases: |
ARCD1, ARP1 |
Location: |
Chr.6:41053201-41064891 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC2 | NM_006789.3 | NP_006780.1 | ||
| AF161698.1 | AAD45360.1 | |||
| AK223461.1 | BAD97181.1 | |||
| AK300689.1 | ||||
| AK308197.1 | ||||
| AK310006.1 | ||||
| AK313288.1 | ||||
| BC047767.2 | AAH47767.1 | |||
| BC069688.1 | AAH69688.1 | |||
| BC069764.1 | AAH69764.1 | |||
| BI438931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526577 | Chr.6:41037763 - 41061603 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map