Assay Details
Target Gene Details
Entrez Gene ID: | 26097 |
Gene Name: | chromatin target of PRMT1 |
Gene Aliases: |
C1orf77, FL-SRAG, FOP, SRAG, SRAG-3, SRAG-5, pp7704 |
Location: |
Chr.1:153633982-153646306 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHTOP | NM_001206612.1 | 6 | 1319 | NP_001193541.1 |
| NM_001244664.1 | 4 | 994 | NP_001231593.1 | |
| NM_001317077.1 | 5 | 1178 | NP_001304006.1 | |
| NM_015607.3 | 6 | 1316 | NP_056422.2 | |
| XM_017000993.1 | 5 | 1140 | XP_016856482.1 | |
| AF261137.1 | AAG44673.1 | |||
| AF318362.1 | 6 | 1225 | AAL55869.1 | |
| AK095230.1 | ||||
| AK097870.1 | ||||
| AL050260.1 | 1 | 987 | CAB43362.2 | |
| AL512704.1 | CAC21650.2 | |||
| BC002733.2 | AAH02733.1 | |||
| BC059949.1 | AAH59949.1 | |||
| BC070027.1 | 1 | 558 | AAH70027.1 | |
| BC108721.1 | 5 | 954 | AAI08722.1 | |
| BC120961.1 | ||||
| BC120962.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv582n54 | Chr.1:153639657 - 153750561 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map