Assay Details
Target Gene Details
Entrez Gene ID: | 148252 |
Gene Name: | DIRAS family GTPase 1 |
Gene Aliases: |
Di-Ras1, GBTS1, RIG |
Location: |
Chr.19:2714567-2721392 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIRAS1 | NM_145173.3 | 2 | 2733 | NP_660156.1 |
| XM_011527713.2 | 2 | 2898 | XP_011526015.1 | |
| AK124445.1 | 1 | 2044 | ||
| AK124672.1 | 1 | 1496 | ||
| AL833958.1 | 1 | 1522 | ||
| BC030660.1 | 2 | 2712 | AAH30660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819697 | Chr.19:2715088 - 2716130 on Build GRCh38 | Loss |
|
| nsv828399 | Chr.19:2714966 - 2724570 on Build GRCh38 | Gain |
|
| nsv578398 | Chr.19:2701508 - 2717882 on Build GRCh38 | Gain |
|
| nsv509714 | Chr.19:2519629 - 2794403 on Build GRCh38 | Insertion |
|
| nsv470109 | Chr.19:2557612 - 2719369 on Build GRCh38 | Loss |
|
| nsv828397 | Chr.19:2688987 - 2800506 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs372966956] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map