Assay Details
Target Gene Details
Entrez Gene ID: | 56832 |
Gene Name: | interferon kappa |
Gene Aliases: |
IFNT1, INFE1 |
Location: |
Chr.9:27524314-27526498 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFNK | NM_020124.2 | 1 | 529 | NP_064509.2 |
| AF146759.1 | 1 | 551 | AAF67468.1 | |
| AF315688.1 | 1 | 540 | AAK01623.1 | |
| AY358855.1 | 1 | 507 | AAQ89214.1 |
Target Gene Details
Entrez Gene ID: | 79817 |
Gene Name: | MOB kinase activator 3B |
Gene Aliases: |
C9orf35, MOB1D, MOBKL2B |
Location: |
Chr.9:27325209-27529852 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB3B | NM_024761.4 | NP_079037.3 | ||
| AK023266.1 | BAB14497.1 | |||
| AL832572.2 | CAD89934.1 | |||
| AU126938.1 | ||||
| BC033027.1 | AAH33027.1 | |||
| DB266074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map