Assay Details
Target Gene Details
Entrez Gene ID: | 83401 |
Gene Name: | ELOVL fatty acid elongase 3 |
Gene Aliases: |
CIG-30, CIG30 |
Location: |
Chr.10:102224767-102229589 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL3 | NM_152310.2 | 3 | 623 | NP_689523.1 |
| XM_011540245.2 | 4 | 1613 | XP_011538547.1 | |
| BC034344.1 | 3 | 515 | AAH34344.1 |
More Information
Set Membership: |
|

Genomic Map