Assay Details
Target Gene Details
Entrez Gene ID: | 84932 |
Gene Name: | RAB2B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.14:21459020-21476973 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB2B | NM_001163380.1 | 7 | 2037 | NP_001156852.1 |
| NM_032846.3 | 8 | 2110 | NP_116235.2 | |
| NR_028074.1 | 8 | 2066 | ||
| XM_017021710.1 | 8 | 2138 | XP_016877199.1 | |
| XM_017021711.1 | 7 | 1935 | XP_016877200.1 | |
| XM_017021712.1 | 7 | 1878 | XP_016877201.1 | |
| AF468652.1 | 9 | 1498 | AAN86142.1 | |
| AK027730.1 | 8 | 935 | BAB55326.1 | |
| BC020839.1 | 9 | 1494 | AAH20839.1 | |
| BC066366.1 | 9 | 1654 | AAH66366.1 | |
| BX647601.1 | 9 | 1176 | CAI46103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map