Assay Details
Target Gene Details
Entrez Gene ID: | 63893 |
Gene Name: | ubiquitin conjugating enzyme E2 O |
Gene Aliases: |
E2-230K |
Location: |
Chr.17:76389532-76453206 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2O | NM_022066.3 | 18 | 3762 | NP_071349.3 |
| XM_005257567.1 | 18 | 3749 | XP_005257624.1 | |
| XM_017024934.1 | 18 | 3674 | XP_016880423.1 | |
| XM_017024935.1 | 18 | 3671 | XP_016880424.1 | |
| AB051521.1 | 18 | 3762 | ||
| AK000926.1 | ||||
| AK022940.1 | 5 | 1260 | BAB14320.1 | |
| AK024657.1 | 3 | 761 | BAB14948.1 | |
| AK025999.1 | 10 | 2241 | BAB15313.1 | |
| AK225412.1 | 10 | 2241 | ||
| AL832432.1 | 11 | 2687 | CAH10644.1 | |
| BC004525.2 | 3 | 577 | AAH04525.2 | |
| BC022237.1 | 5 | 1280 | AAH22237.1 | |
| BC025977.1 | 5 | 1161 | AAH25977.1 | |
| BC036820.1 | 13 | 2898 | AAH36820.1 | |
| BC051868.2 | 10 | 2105 | AAH51868.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2145 | Chr.17:76363105 - 76408339 on Build GRCh38 | Deletion |
|
| nsv833547 | Chr.17:76208499 - 76399227 on Build GRCh38 | Loss |
|
| nsv833549 | Chr.17:76365444 - 76527890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map