Assay Details
Target Gene Details
Entrez Gene ID: | 60509 |
Gene Name: | ATP/GTP binding protein like 5 |
Gene Aliases: |
CCP5, RP75 |
Location: |
Chr.2:27051423-27070622 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL5 | NM_001035507.2 | NP_001030584.1 | ||
| NM_021831.5 | NP_068603.4 | |||
| NR_104246.1 | ||||
| XM_005264477.2 | XP_005264534.1 | |||
| XM_011533011.2 | XP_011531313.1 | |||
| XM_011533012.2 | XP_011531314.1 | |||
| XM_011533013.2 | XP_011531315.1 | |||
| AK023398.1 | BAB14560.1 | |||
| AK025492.1 | BAB15151.1 | |||
| AK302091.1 | ||||
| AL833844.1 | CAD38704.1 | |||
| BC007415.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss |
|
| nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain |
|
| nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map