Assay Details
Target Gene Details
Entrez Gene ID: | 284427 |
Gene Name: | solute carrier family 25 member 41 |
Gene Aliases: |
APC4 |
Location: |
Chr.19:6426037-6436267 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A41 | NM_001321298.1 | NP_001308227.1 | ||
| NM_173637.3 | NP_775908.2 | |||
| NR_135612.1 | ||||
| XM_011527926.1 | XP_011526228.1 | |||
| AK097761.1 | BAC05163.1 | |||
| BC031671.1 | AAH31671.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map