Assay Details
Target Gene Details
Entrez Gene ID: | 55893 |
Gene Name: | zinc finger protein 395 |
Gene Aliases: |
HDBP-2, HDBP2, HDRF-2, PBF, PRF-1, PRF1, Si-1-8-14 |
Location: |
Chr.8:28345585-28386460 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF395 | NM_018660.2 | 9 | 1501 | NP_061130.1 |
| AB044750.1 | 8 | 1324 | BAA96783.1 | |
| AB044752.1 | BAA96785.1 | |||
| AB073627.1 | 9 | 1507 | BAD29734.1 | |
| AF263928.1 | 10 | 1711 | AAF73463.1 | |
| AK002050.1 | 9 | 1426 | BAA92056.1 | |
| AK021850.1 | 2 | 1360 | ||
| AK023418.1 | 9 | 1501 | BAB14571.1 | |
| AK098243.1 | 15 | 2384 | ||
| BC001237.1 | 9 | 1463 | AAH01237.1 | |
| BG249131.1 | 1 | 55 | ||
| BG759384.1 | 3 | 225 | ||
| BI917232.1 | 2 | 118 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv456n21 | Chr.8:28321496 - 28467894 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75444725] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map