Assay Details
Target Gene Details
Entrez Gene ID: | 2064 |
Gene Name: | erb-b2 receptor tyrosine kinase 2 |
Gene Aliases: |
CD340, HER-2, HER-2/neu, HER2, MLN 19, NEU, NGL, TKR1 |
Location: |
Chr.17:39688084-39728662 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERBB2 | NM_001005862.2 | 30 | 4690 | NP_001005862.1 |
| NM_001289936.1 | 31 | 4741 | NP_001276865.1 | |
| NM_001289937.1 | 26 | 4212 | NP_001276866.1 | |
| NM_004448.3 | 27 | 4465 | NP_004439.2 | |
| NR_110535.1 | 27 | 4528 | ||
| BM678576.1 | 1 | 200 | ||
| M11730.1 | 27 | 4354 | AAA75493.1 | |
| X03363.1 | 27 | 4378 | CAA27060.1 |
Target Gene Details
Entrez Gene ID: | 84299 |
Gene Name: | migration and invasion enhancer 1 |
Gene Aliases: |
C17orf37, C35, ORB3, RDX12, XTP4 |
Location: |
Chr.17:39728500-39730563 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIEN1 | NM_032339.4 | NP_115715.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056234 | Chr.17:39706352 - 39753735 on Build GRCh38 | Gain |
|
| nsv2048 | Chr.17:39718682 - 39742588 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map