Assay Details
Target Gene Details
Entrez Gene ID: | 2288 |
Gene Name: | FK506 binding protein 4 |
Gene Aliases: |
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52 |
Location: |
Chr.12:2794942-2805423 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP4 | NM_002014.3 | NP_002005.1 | ||
| XM_011520929.2 | XP_011519231.1 | |||
| AK313918.1 | ||||
| BC001786.1 | AAH01786.1 | |||
| BC007924.2 | AAH07924.1 | |||
| M88279.1 | AAA36111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047874 | Chr.12:2577599 - 2807322 on Build GRCh38 | Gain |
|
| esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain |
|
| esv3628322 | Chr.12:2796908 - 2837171 on Build GRCh38 | Gain |
|
| nsv826194 | Chr.12:2777359 - 2817185 on Build GRCh38 | Gain |
|
| dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map