Assay Details
Target Gene Details
Entrez Gene ID: | 26027 |
Gene Name: | acyl-CoA thioesterase 11 |
Gene Aliases: |
BFIT, STARD14, THEA, THEM1 |
Location: |
Chr.1:54548134-54634744 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT11 | NM_015547.3 | 1 | 121 | NP_056362.1 |
| NM_147161.3 | 1 | 121 | NP_671517.1 | |
| AB014607.1 | ||||
| AF416921.1 | AAL40937.1 | |||
| AF416922.1 | AAL40938.1 | |||
| AK023937.1 | BAB14734.1 | |||
| AK091112.1 | ||||
| AK223570.1 | BAD97290.1 | |||
| BC001517.2 | AAH01517.1 | |||
| BC093844.1 | AAH93844.1 | |||
| BC093846.1 | AAH93846.1 | |||
| DA121227.1 | 1 | 121 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461561 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| nsv546267 | Chr.1:54518434 - 54604313 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map