Assay Details
Target Gene Details
Entrez Gene ID: | 100500827 |
Gene Name: | microRNA 3614 |
Gene Aliases: |
mir-3614 |
Location: |
Chr.17:56891270-56891355 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR3614 | NR_037408.1 |
Target Gene Details
Entrez Gene ID: | 7706 |
Gene Name: | tripartite motif containing 25 |
Gene Aliases: |
EFP, RNF147, Z147, ZNF147 |
Location: |
Chr.17:56887909-56914048 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM25 | NM_005082.4 | 9 | 2357 | NP_005073.2 |
| AB208994.1 | BAD92231.1 | |||
| BC016924.1 | AAH16924.1 | |||
| BC042541.1 | AAH42541.1 | |||
| BX538220.1 | ||||
| D21205.1 | BAA04747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32884 | Chr.17:56683534 - 57002132 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map