Assay Details
Target Gene Details
Entrez Gene ID: | 79887 |
Gene Name: | phospholipase B domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.12:14503663-14567857 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLBD1 | NM_024829.5 | NP_079105.4 | ||
| AK026315.1 | BAB15442.1 | |||
| AK225458.1 | ||||
| AK290914.1 | ||||
| BC000909.2 | AAH00909.2 | |||
| BC063561.1 | AAH63561.1 | |||
| CD366142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557608 | Chr.12:14469068 - 14535710 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map