Assay Details
Target Gene Details
Entrez Gene ID: | 64645 |
Gene Name: | major facilitator superfamily domain containing 14A |
Gene Aliases: |
HIAT1 |
Location: |
Chr.1:100038078-100083377 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD14A | NM_033055.2 | NP_149044.2 | ||
| XM_005271118.1 | XP_005271175.1 | |||
| XM_011541957.1 | XP_011540259.1 | |||
| XM_017002084.1 | XP_016857573.1 | |||
| AF427492.1 | AAL25115.1 | |||
| AK057172.1 | BAB71375.1 | |||
| AK096669.1 | BAC04836.1 | |||
| BC064409.1 | AAH64409.1 | |||
| BC107768.1 | AAI07769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
| nsv546888 | Chr.1:99954311 - 100084889 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78415949] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map