Assay Details
Target Gene Details
Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5SL | NM_001167867.1 | 6 | 1040 | NP_001161339.1 |
| NM_001167868.1 | 5 | 877 | NP_001161340.1 | |
| NM_001167869.1 | 5 | 831 | NP_001161341.1 | |
| NM_001167870.1 | 4 | 750 | NP_001161342.1 | |
| NM_001167871.1 | 5 | 913 | NP_001161343.1 | |
| NM_001320838.1 | 5 | 794 | NP_001307767.1 | |
| NM_001320840.1 | 5 | 959 | NP_001307769.1 | |
| NM_001320841.1 | 4 | 796 | NP_001307770.1 | |
| NM_018035.2 | 6 | 994 | NP_060505.2 | |
| NR_030765.1 | 6 | 977 | ||
| NR_135476.1 | 5 | 845 | ||
| NR_135477.1 | 6 | 991 | ||
| NR_135478.1 | 6 | 1008 | ||
| XM_011527065.2 | 6 | 1045 | XP_011525367.1 | |
| AK001103.1 | 6 | 961 | BAA91503.1 | |
| AK123631.1 | 3 | 1586 | ||
| AK293132.1 | 5 | 840 | ||
| AK297698.1 | 5 | 801 | ||
| AK298197.1 | 5 | 912 | ||
| AK301940.1 | 6 | 1003 | ||
| AK316313.1 | 5 | 886 | ||
| BC013323.1 | 4 | 726 | AAH13323.1 |

Genomic Map