Assay Details
Target Gene Details
Entrez Gene ID: | 2135 |
Gene Name: | exostosin like glycosyltransferase 2 |
Gene Aliases: |
EXTR2 |
Location: |
Chr.1:100872372-100895411 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXTL2 | NM_001033025.2 | 3 | 883 | NP_001028197.1 |
| NM_001261440.1 | 3 | 883 | NP_001248369.1 | |
| NM_001261441.1 | 4 | 907 | NP_001248370.1 | |
| NM_001261442.1 | 3 | 919 | NP_001248371.1 | |
| NM_001439.3 | 3 | 919 | NP_001430.1 | |
| NR_048570.1 | 4 | 1014 | ||
| XM_005270621.1 | 4 | 749 | XP_005270678.1 | |
| XM_011540994.2 | 4 | 1175 | XP_011539296.1 | |
| XM_011540995.2 | 5 | 1192 | XP_011539297.1 | |
| XM_011540996.2 | 4 | 397 | XP_011539298.1 | |
| XM_017000651.1 | 3 | 523 | XP_016856140.1 | |
| AB009284.1 | 3 | 552 | BAA24081.1 | |
| AF000416.1 | 3 | 552 | AAC02898.1 | |
| AK096914.1 | 3 | 516 | BAC04893.1 | |
| AK298124.1 | 4 | 667 | ||
| AK312895.1 | 3 | 488 | ||
| AU130571.1 | 4 | 392 | ||
| BC014910.1 | 3 | 439 | ||
| BC036015.1 | 3 | 529 | AAH36015.1 | |
| BC045681.1 | 3 | 1702 | AAH45681.1 | |
| BP202626.1 | 1 | 220 | ||
| BX103291.1 | 3 | 552 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map