Assay Details
Target Gene Details
Entrez Gene ID: | 57467 |
Gene Name: | hedgehog acyltransferase-like |
Gene Aliases: |
C3orf3, GUP1, MBOAT3, MSTP002, OACT3 |
Location: |
Chr.3:42692663-42702849 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHATL | NM_020707.3 | NP_065758.3 | ||
| NR_027753.1 | ||||
| XM_006713274.2 | XP_006713337.1 | |||
| XM_006713275.2 | XP_006713338.1 | |||
| XM_011533969.2 | XP_011532271.1 | |||
| XM_011533970.2 | XP_011532272.1 | |||
| XM_017006935.1 | XP_016862424.1 | |||
| XM_017006936.1 | XP_016862425.1 | |||
| XM_017006937.1 | XP_016862426.1 | |||
| XM_017006938.1 | XP_016862427.1 | |||
| AB042554.1 | BAB17758.1 | |||
| BC022529.1 | AAH22529.1 | |||
| BI544867.1 | ||||
| BI596471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954472 | Chr.3:42683809 - 42713908 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map