Assay Details
Target Gene Details
Entrez Gene ID: | 8728 |
Gene Name: | ADAM metallopeptidase domain 19 |
Gene Aliases: |
FKSG34, MADDAM, MLTNB |
Location: |
Chr.5:157477304-157575823 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM19 | NM_033274.4 | 23 | 6376 | NP_150377.1 |
| XM_011534682.2 | 16 | 7132 | XP_011532984.1 | |
| XM_017010009.1 | 16 | 5593 | XP_016865498.1 | |
| AK091464.1 | ||||
| AK093535.1 | 1 | 2030 | ||
| AL049415.1 | 1 | 1087 | ||
| BC024214.2 | 12 | 1694 | AAH24214.2 | |
| BC033132.1 | 12 | 1694 | AAH33132.1 | |
| Y13786.2 | 23 | 6326 | CAC20585.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462497 | Chr.5:157462089 - 157493903 on Build GRCh38 | Loss |
|
| nsv830526 | Chr.5:157325531 - 157527933 on Build GRCh38 | Loss |
|
| nsv5089 | Chr.5:157454904 - 157490416 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map