Assay Details
Target Gene Details
Entrez Gene ID: | 56147 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
PCDH-ALPHA1 |
Location: |
Chr.5:140786136-141012344 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA1 | NM_018900.3 | 1 | 201 | NP_061723.1 |
| NM_031410.2 | 1 | 201 | NP_113598.1 | |
| NM_031411.2 | 1 | 201 | NP_113599.1 | |
| AF152305.1 | AAD43699.1 | |||
| AF152479.1 | AAD43740.1 | |||
| AF169695.1 | AAF89692.1 | |||
| DA330326.1 | 1 | 201 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv830508 | Chr.5:140675388 - 140869073 on Build GRCh38 | Loss |
|
| dgv970n67 | Chr.5:140784829 - 140843644 on Build GRCh38 | Gain |
|
| nsv964946 | Chr.5:140786102 - 140786602 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map