Assay Details
Target Gene Details
Entrez Gene ID: | 729220 |
Gene Name: | uncharacterized LOC729220 |
Gene Aliases: |
- |
Location: |
Chr.17:49845910-49848837 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ45513 | NM_001242791.1 | 3 | 1170 | NP_001229720.1 |
| AK127421.1 | 3 | 1170 | BAC86972.1 |
Target Gene Details
Entrez Gene ID: | 255061 |
Gene Name: | tachykinin 4 (hemokinin) |
Gene Aliases: |
EK, HK-1, HK1, PPT-C |
Location: |
Chr.17:49838309-49848017 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAC4 | NM_001077503.1 | NP_001070971.1 | ||
| NM_001077504.1 | NP_001070972.1 | |||
| NM_001077505.1 | NP_001070973.1 | |||
| NM_001077506.1 | NP_001070974.1 | |||
| NM_170685.2 | NP_733786.2 | |||
| AF515828.1 | AAP30874.1 | |||
| AF515829.1 | AAP30875.1 | |||
| AF515830.1 | AAP30876.1 | |||
| AF515831.1 | AAP30877.1 | |||
| AF521560.1 | AAN77128.1 | |||
| AY471574.1 | AAS46596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828037 | Chr.17:49845899 - 49847886 on Build GRCh38 | Gain |
|
| nsv821118 | Chr.17:49845825 - 49847886 on Build GRCh38 | Duplication |
|
| esv25474 | Chr.17:49846051 - 49847886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map