Assay Details
Target Gene Details
Entrez Gene ID: | 57696 |
Gene Name: | DEAD-box helicase 55 |
Gene Aliases: |
- |
Location: |
Chr.12:123602077-123620943 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX55 | NM_020936.2 | NP_065987.1 | ||
| NR_135104.1 | ||||
| NR_135105.1 | ||||
| XM_017019710.1 | XP_016875199.1 | |||
| XM_017019711.1 | XP_016875200.1 | |||
| XM_017019712.1 | XP_016875201.1 | |||
| XM_017019713.1 | XP_016875202.1 | |||
| XM_017019714.1 | XP_016875203.1 | |||
| XM_017019715.1 | XP_016875204.1 | |||
| XM_017019716.1 | XP_016875205.1 | |||
| XM_017019717.1 | XP_016875206.1 | |||
| XM_017019718.1 | XP_016875207.1 | |||
| XM_017019719.1 | XP_016875208.1 | |||
| XM_017019720.1 | XP_016875209.1 | |||
| AB046815.1 | BAB13421.1 | |||
| AK131442.1 | ||||
| BC030020.2 | AAH30020.1 | |||
| BC035911.1 | AAH35911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1566n100 | Chr.12:123610211 - 123711309 on Build GRCh38 | Gain |
|
| dgv524e212 | Chr.12:123614065 - 123625575 on Build GRCh38 | Loss |
|
| nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map