Assay Details
Target Gene Details
Entrez Gene ID: | 105376488 |
Gene Name: | uncharacterized LOC105376488 |
Gene Aliases: |
- |
Location: |
Chr.10:33334437-33385325 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376488 | XM_017017010.1 | XP_016872499.1 |
Target Gene Details
Entrez Gene ID: | 8829 |
Gene Name: | neuropilin 1 |
Gene Aliases: |
BDCA4, CD304, NP1, NRP, VEGF165R |
Location: |
Chr.10:33177491-33334905 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRP1 | NM_001024628.2 | 1 | 309 | NP_001019799.1 |
| NM_001024629.2 | 1 | 309 | NP_001019800.1 | |
| NM_001244972.1 | 1 | 309 | NP_001231901.1 | |
| NM_001244973.1 | 1 | 309 | NP_001231902.1 | |
| NM_003873.5 | 1 | 309 | NP_003864.4 | |
| NR_045259.1 | 1 | 309 | ||
| XM_006717521.1 | 1 | 83 | XP_006717584.1 | |
| XM_006717522.1 | 1 | 83 | XP_006717585.1 | |
| XM_006717523.2 | 1 | 83 | XP_006717586.1 | |
| XM_006717524.1 | 1 | 83 | XP_006717587.1 | |
| XM_006717525.1 | 1 | 83 | XP_006717588.1 | |
| XM_006717526.1 | 1 | 83 | XP_006717589.1 | |
| XM_011519755.2 | 1 | 83 | XP_011518057.1 | |
| XM_011519756.1 | 1 | 83 | XP_011518058.1 | |
| XM_017016865.1 | 1 | 83 | XP_016872354.1 | |
| XM_017016866.1 | 1 | 85 | XP_016872355.1 | |
| AK292822.1 | ||||
| AL698999.1 | ||||
| AL833608.1 | CAH10397.1 | |||
| BC007533.1 | AAH07533.1 | |||
| BX648025.1 | 2 | 334 | CAH10373.1 | |
| BX648250.1 | 1 | 279 | ||
| CR749333.1 | 1 | 310 | CAH18187.1 | |
| DA702852.1 | 2 | 296 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831829 | Chr.10:33178304 - 33344113 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map