Assay Details
Target Gene Details
Entrez Gene ID: | 10873 |
Gene Name: | malic enzyme 3 |
Gene Aliases: |
NADP-ME |
Location: |
Chr.11:86441108-86674745 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ME3 | NM_001014811.1 | NP_001014811.1 | ||
| NM_001161586.1 | NP_001155058.1 | |||
| NM_006680.2 | NP_006671.2 | |||
| XM_005273717.1 | XP_005273774.1 | |||
| XM_017017136.1 | XP_016872625.1 | |||
| XM_017017137.1 | XP_016872626.1 | |||
| XM_017017138.1 | XP_016872627.1 | |||
| AI560417.1 | ||||
| AI948779.1 | ||||
| AK313695.1 | ||||
| BC022472.2 | AAH22472.1 | |||
| BQ882896.1 | ||||
| CB306059.1 | ||||
| X79440.1 | CAA55956.1 |
More Information
Set Membership: |
|

Genomic Map