Assay Details
Target Gene Details
Entrez Gene ID: | 8284 |
Gene Name: | lysine demethylase 5D |
Gene Aliases: |
HY, HYA, JARID1D, SMCY |
Location: |
Chr.Y:19692495-19745347 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM5D | NM_001146705.1 | 28 | 4799 | NP_001140177.1 |
| NM_001146706.1 | 26 | 4535 | NP_001140178.1 | |
| NM_004653.4 | 27 | 4706 | NP_004644.2 | |
| XM_005262560.2 | 26 | 4979 | XP_005262617.1 | |
| XM_005262561.2 | 27 | 4883 | XP_005262618.1 | |
| XM_011531468.2 | 27 | 5036 | XP_011529770.1 | |
| AK095923.1 | 11 | 1909 | ||
| AK127269.1 | 15 | 4623 | ||
| BC132721.1 | 27 | 4629 | ||
| BC144102.1 | 28 | 4722 | ||
| BC146767.1 | 26 | 4357 | ||
| BX648643.1 | 12 | 3169 | ||
| D87072.1 | 26 | 4357 | ||
| U52191.1 | 27 | 4693 | AAC50806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10025 | Chr.Y:18865177 - 20859250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map